This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04764879_g1
See other MOGS GE Assays ›
Gene Symbol:
MOGS
Gene Name:
mannosyl-oligosaccharide glucosidase
Gene Aliases:
-
Chromosome Location:
Chr.11: 10008608 - 10012385 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04764879_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MOGS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005686325.3 XP_005686382.3 3 - 4 942 78
GenBank mRNA KA350564.1 - 1989 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

glucosidase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline