This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04765523_m1
See other SPIB GE Assays ›
Gene Symbol:
SPIB
Gene Name:
Spi-B transcription factor
Gene Aliases:
-
Chromosome Location:
Chr.18: 57587350 - 57593202 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04765523_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SPIB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018062882.1 XP_017918371.1 4 - 5 719 87
XM_018062883.1 XP_017918372.1 5 - 6 488 87
XM_018062884.1 XP_017918373.1 2 - 3 218 87
GenBank mRNA JO581598.1 - 1 - 2 2209 87

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

winged helix/forkhead transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0