This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04766032_m1
See other DLC1 GE Assays ›
Gene Symbol:
DLC1
Gene Name:
DLC1 Rho GTPase activating protein
Gene Aliases:
-
Chromosome Location:
Chr.27: 21679680 - 22113391 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04766032_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DLC1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005698779.3 XP_005698836.2 15 - 16 4504 61
XM_005698780.3 XP_005698837.2 11 - 12 2958 61
XM_005698781.3 XP_005698838.1 11 - 12 2883 61
XM_018041886.1 XP_017897375.1 15 - 16 4360 61
XM_018041888.1 XP_017897377.1 11 - 12 3148 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

G-protein modulator protein-binding activity modulator GTPase-activating protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0