This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04770098_g1
See other HYI GE Assays ›
Gene Symbol:
HYI
Gene Name:
hydroxypyruvate isomerase (putative)
Gene Aliases:
-
Chromosome Location:
Chr.3: 18187335 - 18190215 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04770098_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HYI
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005709577.3 XP_005709634.3 1 - 2 335 60
XM_018042897.1 XP_017898386.1 1 - 2 335 60
XM_018042904.1 XP_017898393.1 1 - 2 335 60
GenBank mRNA JT664823.1 - 1 - 2 272 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

isomerase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline