This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04771909_m1
See other TVP23A GE Assays ›
Gene Symbol:
TVP23A
Gene Name:
trans-golgi network vesicle protein 23 homolog A
Gene Aliases:
-
Chromosome Location:
Chr.25: 9345583 - 9376674 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04771909_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TVP23A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018040870.1 XP_017896359.1 1 - 2 952 67
XM_018040871.1 XP_017896360.1 1 - 2 952 67
XM_018040872.1 XP_017896361.1 3 - 4 401 67
XM_018040873.1 XP_017896362.1 3 - 4 397 67
GenBank mRNA JO641257.2 - 5 - 6 1545 67
JO641258.2 - 11 - 12 1409 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zllbd:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0