This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04776046_m1
See other BOLL GE Assays ›
Gene Symbol:
BOLL
Gene Name:
boule homolog, RNA binding protein
Gene Aliases:
boule
Chromosome Location:
Chr.2: 49662972 - 49727087 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04776046_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BOLL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005676311.2 XP_005676368.1 10 - 11 976 74
XM_005676312.2 XP_005676369.1 10 - 11 940 74
XM_005676313.2 XP_005676370.1 9 - 10 904 74
XM_005676314.3 XP_005676371.1 8 - 9 808 74
GenBank mRNA JQ946560.1 - 8 - 9 744 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein nucleic acid metabolism protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gjvvj:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0