This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04777664_m1
See other PDE12 GE Assays ›
Gene Symbol:
PDE12
Gene Name:
phosphodiesterase 12
Gene Aliases:
-
Chromosome Location:
Chr.22: 43569317 - 43576106 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04777664_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PDE12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005695819.3 XP_005695876.3 1 - 2 2972 96
GenBank mRNA JO431502.2 - 2 - 3 732 96

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

mRNA polyadenylation factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-s8bwd:80/100.66.79.198:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline