This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04779503_m1
See other PTPN2 GE Assays ›
Gene Symbol:
PTPN2
Gene Name:
protein tyrosine phosphatase non-receptor type 2
Gene Aliases:
-
Chromosome Location:
Chr.24: 43436685 - 43502641 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04779503_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PTPN2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018039609.1 XP_017895098.1 3 - 4 443 67
XM_018039610.1 XP_017895099.1 3 - 4 443 67
XM_018039611.1 XP_017895100.1 3 - 4 448 67
GenBank mRNA JO579775.1 - 3 - 4 368 67
JO579776.1 - 3 - 4 368 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

protein phosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0