This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04780654_m1
See other LDAH GE Assays ›
Gene Symbol:
LDAH
Gene Name:
lipid droplet associated hydrolase
Gene Aliases:
C11H2orf43
Chromosome Location:
Chr.11: 77798690 - 77896693 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04780654_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LDAH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005686967.3 XP_005687024.2 2 - 3 262 72
XM_005686968.3 XP_005687025.2 2 - 3 668 72
XM_005686969.3 XP_005687026.2 2 - 3 262 72
XM_013967840.2 XP_013823294.2 2 - 3 262 72
XM_018055591.1 XP_017911080.1 2 - 3 262 72
GenBank mRNA JO429616.2 - 5 - 6 1931 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

lipase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline