This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04781298_g1
See other STARD5 GE Assays ›
Gene Symbol:
STARD5
Gene Name:
StAR related lipid transfer domain containing 5
Gene Aliases:
-
Chromosome Location:
Chr.21: 26738956 - 26747553 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04781298_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: STARD5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005695076.3 XP_005695133.1 2 - 3 261 73
XM_013973085.2 XP_013828539.1 2 - 3 186 73
XM_013973086.2 XP_013828540.1 2 - 3 262 73
GenBank mRNA JO397626.2 - 2 - 3 246 73
KA342468.1 - 2 - 3 221 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0