This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04791252_g1
See other PGGHG GE Assays ›
Gene Symbol:
PGGHG
Gene Name:
protein-glucosylgalactosylhydroxylysine glucosidase
Gene Aliases:
-
Chromosome Location:
Chr.29: 46035920 - 46042087 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04791252_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PGGHG
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018042970.1 XP_017898459.1 10 - 11 2105 63
XM_018042971.1 XP_017898460.1 10 - 11 2082 63
XM_018042972.1 XP_017898461.1 10 - 11 2037 63
GenBank mRNA KA314974.1 - 7 - 8 758 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

glycosidase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0