This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04800141_g1
See other HSD11B1L GE Assays ›
Gene Symbol:
HSD11B1L
Gene Name:
hydroxysteroid 11-beta dehydrogenase 1 like
Gene Aliases:
-
Chromosome Location:
Chr.7: 91907627 - 91912578 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04800141_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HSD11B1L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005682511.2 XP_005682568.1 5 - 6 682 65
XM_005682512.3 XP_005682569.1 5 - 6 651 65
XM_005682513.3 XP_005682570.1 4 - 5 535 65
XM_013965251.2 XP_013820705.1 4 - 5 2322 65
XM_013965252.2 XP_013820706.1 4 - 5 2322 65
XM_013965253.2 XP_013820707.1 3 - 4 2206 65
XM_013965254.1 XP_013820708.1 3 - 4 361 65
XM_013965255.1 XP_013820709.1 2 - 3 245 65
XM_018050910.1 XP_017906399.1 4 - 5 566 65

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_013965254.1, XM_013965255.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0