This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04802065_m1
See other PPP2R2C GE Assays ›
Gene Symbol:
PPP2R2C
Gene Name:
protein phosphatase 2 regulatory subunit Bgamma
Gene Aliases:
-
Chromosome Location:
Chr.6: 103089844 - 103254694 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04802065_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPP2R2C
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_013964782.2 XP_013820236.2 8 - 9 1281 63
XM_018049709.1 XP_017905198.1 8 - 9 1049 63
GenBank mRNA JO631750.2 - 8 - 9 1433 63
JO631751.2 - 8 - 9 1433 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

protein phosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline