This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04804569_m1
See other TMPRSS13 GE Assays ›
Gene Symbol:
TMPRSS13
Gene Name:
transmembrane serine protease 13
Gene Aliases:
-
Chromosome Location:
Chr.15: 53734368 - 53767559 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04804569_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMPRSS13
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005689500.3 XP_005689557.2 8 - 9 894 55
XM_013969565.2 XP_013825019.2 8 - 9 927 55
XM_013969566.2 XP_013825020.2 7 - 8 888 55
XM_013969567.2 XP_013825021.2 7 - 8 855 55
GenBank mRNA KA304837.1 - 7 - 8 936 55

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

serine protease

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-dd86746ff-lmgjf:80/100.66.77.199:80.
git-commit: 04e44c58f20fafe66519586ffe6db5d8a90e52ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.53.1-Offline