Assay Details
| Gene Symbol: CDCA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064769.1 | XP_017920258.1 | 7 - 8 | 1158 | 66 | |
| XM_018064778.1 | XP_017920267.1 | 6 - 7 | 903 | 66 | ||
| GenBank mRNA | JO609251.2 | - | 3 - 4 | 1545 | 66 | |
More Information
Set Membership: |
|

CDCA7
Genomic Map