This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04807761_mH
See other VTA1 GE Assays ›
Gene Symbol:
VTA1
Gene Name:
vesicle trafficking 1
Gene Aliases:
-
Chromosome Location:
Chr.9: 66664722 - 66737541 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04807761_mH
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: VTA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005684854.3 XP_005684911.1 1 - 2 166 99
XM_005684855.3 XP_005684912.1 1 - 2 176 99
GenBank mRNA JO645609.2 - 7 - 8 1271 99
JO645610.2 - 7 - 8 1268 99

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

membrane traffic protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0