This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj00533885_g1
See other PTPN11 GE Assays ›
Gene Symbol:
PTPN11
Gene Name:
protein tyrosine phosphatase, non-receptor type 11
Gene Aliases:
-
Chromosome Location:
Chr.9: 102716174 - 102819938 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj00533885_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PTPN11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002753025.2 XP_002753071.1 8 - 9 1305 79
XM_009004695.2 XP_009002943.1 8 - 9 1072 79
XM_009004696.2 XP_009002944.1 8 - 9 1070 79
XM_009004697.2 XP_009002945.1 8 - 9 1146 79
XM_017976389.1 XP_017831878.1 8 - 9 1071 79
XM_017976390.1 XP_017831879.1 8 - 9 1133 79

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

protein phosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0