This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj00894794_m1
See other MIER1 GE Assays ›
Gene Symbol:
MIER1
Gene Name:
MIER1 transcriptional regulator
Gene Aliases:
-
Chromosome Location:
Chr.7: 102512502 - 102576114 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj00894794_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIER1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_009001440.2 XP_008999688.1 12 - 13 1190 128
XM_009001441.1 XP_008999689.1 12 - 13 1211 128
XM_009001442.2 XP_008999690.1 12 - 13 1309 128
XM_009001443.2 XP_008999691.1 12 - 13 1205 128
XM_009001444.2 XP_008999692.1 10 - 11 1085 128
XM_009001445.2 XP_008999693.1 11 - 12 1103 128
XM_009001446.2 XP_008999694.1 11 - 12 1251 128
XM_009001449.2 XP_008999697.1 12 - 13 1309 128
XM_009001450.2 XP_008999698.1 10 - 11 1096 128
XM_017974821.1 XP_017830310.1 11 - 12 1110 128
XM_017974822.1 XP_017830311.1 11 - 12 1110 128
XM_017974823.1 XP_017830312.1 11 - 12 1110 128
XM_017974824.1 XP_017830313.1 11 - 12 1111 128
XM_017974825.1 XP_017830314.1 12 - 13 1153 128
XM_017974826.1 XP_017830315.1 12 - 13 1211 128
XM_017974827.1 XP_017830316.1 13 - 14 1167 128

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

homeodomain transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0