Assay Details
| Gene Symbol: SNAP25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747359.2 | XP_002747405.1 | 7 - 8 | 901 | 64 | |
| XM_002747361.4 | XP_002747407.1 | 7 - 8 | 791 | 64 | ||
| XM_008995747.2 | XP_008993995.1 | 8 - 9 | 943 | 64 | ||
| XM_017971957.1 | XP_017827446.1 | 8 - 9 | 888 | 64 | ||
More Information
Set Membership: |
|

SNAP25
Genomic Map