Assay Details
| Gene Symbol: MRPS18A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746592.4 | XP_002746638.1 | 3 - 4 | 400 | 71 | |
| XM_008994501.2 | XP_008992749.1 | 3 - 4 | 305 | 71 | ||
More Information
Set Membership: |
|

MRPS18A
Genomic Map