This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj02791973_m1
See other UBE2K GE Assays ›
Gene Symbol:
UBE2K
Gene Name:
ubiquitin conjugating enzyme E2 K
Gene Aliases:
-
Chromosome Location:
Chr.3: 151126082 - 151206359 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj02791973_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UBE2K
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002745900.4 XP_002745946.1 4 - 5 582 116
XM_008993465.2 XP_008991713.1 4 - 5 582 116
XM_017970616.1 XP_017826105.1 3 - 4 485 116

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017970616.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4krp5:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0