This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj02876099_m1
See other ELP4 GE Assays ›
Gene Symbol:
ELP4
Gene Name:
elongator acetyltransferase complex subunit 4
Gene Aliases:
-
Chromosome Location:
Chr.11: 94670043 - 94946765 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj02876099_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ELP4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002755140.3 XP_002755186.1 1 - 2 269 92
XM_009007873.2 XP_009006121.1 1 - 2 270 92
XM_017977987.1 XP_017833476.1 1 - 2 270 92
XM_017977988.1 XP_017833477.1 1 - 2 270 92
XM_017977989.1 XP_017833478.1 1 - 2 270 92
XM_017977990.1 XP_017833479.1 1 - 2 270 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0