Assay Details
| Gene Symbol: GLIS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002742900.3 | XP_002742946.1 | 6 - 7 | 2715 | 95 | |
| XM_017973380.1 | XP_017828869.1 | 5 - 6 | 2409 | 95 | ||
More Information
Set Membership: |
|

GLIS3
Genomic Map