This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj02889511_g1
See other AASDH GE Assays ›
Gene Symbol:
AASDH
Gene Name:
aminoadipate-semialdehyde dehydrogenase
Gene Aliases:
-
Chromosome Location:
Chr.3: 137158866 - 137202028 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj02889511_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AASDH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008993310.2 XP_008991558.1 8 - 9 1662 82
XM_008993311.2 XP_008991559.1 8 - 9 1497 82
XM_008993312.2 XP_008991560.1 8 - 9 1626 82
XM_008993313.2 XP_008991561.1 8 - 9 1661 82
XM_008993314.2 XP_008991562.1 6 - 7 1148 82
XM_008993315.1 XP_008991563.1 6 - 7 1110 82
XM_008993316.1 XP_008991564.1 4 - 5 845 82
XM_008993317.1 XP_008991565.1 5 - 6 1003 82
XM_017970551.1 XP_017826040.1 8 - 9 1493 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

ligase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0