Assay Details
| Gene Symbol: ZNF454 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008989852.2 | XP_008988100.1 | 5 - 6 | 882 | 87 | |
| XM_017969489.1 | XP_017824978.1 | 5 - 6 | 773 | 87 | ||
More Information
Set Membership: |
|

ZNF454
Genomic Map