This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj05919217_gH
See other ZNF41 GE Assays ›
Gene Symbol:
ZNF41
Gene Name:
zinc finger protein 41
Gene Aliases:
-
Chromosome Location:
Chr.X: 46414588 - 46461300 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj05919217_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZNF41
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008989203.2 XP_008987451.1 3 - 4 1014 77
XM_008989204.2 XP_008987452.1 3 - 4 878 77
XM_008989205.2 XP_008987453.1 3 - 4 717 77
XM_017968599.1 XP_017824088.1 2 - 3 2057 77
XM_017968600.1 XP_017824089.1 3 - 4 1030 77
XM_017968601.1 XP_017824090.1 3 - 4 733 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0