This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj05924225_m1
See other OPCML GE Assays ›
Gene Symbol:
OPCML
Gene Name:
opioid binding protein/cell adhesion molecule like
Gene Aliases:
-
Chromosome Location:
Chr.11: 30510447 - 31077231 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj05924225_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: OPCML
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_009007154.2 XP_009005402.1 2 - 3 1169 88
XM_009007155.2 XP_009005403.1 2 - 3 1169 88
XM_009007156.2 XP_009005404.1 2 - 3 1169 88
XM_017978560.1 XP_017834049.1 2 - 3 1169 88
XM_017978561.1 XP_017834050.1 3 - 4 669 88
XM_017978562.1 XP_017834051.1 3 - 4 669 88
XM_017978563.1 XP_017834052.1 2 - 3 1169 88
XM_017978564.1 XP_017834053.1 3 - 4 669 88
XM_017978565.1 XP_017834054.1 2 - 3 1169 88

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

immunoglobulin

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline