This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj05926629_g1
See other TRAPPC4 GE Assays ›
Gene Symbol:
TRAPPC4
Gene Name:
trafficking protein particle complex 4
Gene Aliases:
-
Chromosome Location:
Chr.11: 16660098 - 16664966 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj05926629_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TRAPPC4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002754503.4 XP_002754549.1 4 - 5 889 83
XM_017977601.1 XP_017833090.1 2 - 3 262 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

membrane traffic protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-r7bw6:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0