Assay Details
| Gene Symbol: KIF2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750737.3 | XP_002750783.1 | 3 - 4 | 443 | 80 | |
| XM_002750738.3 | XP_002750784.1 | 3 - 4 | 468 | 80 | ||
| XM_009001078.2 | XP_008999326.1 | 3 - 4 | 440 | 80 | ||
| XM_017974654.1 | XP_017830143.1 | 3 - 4 | 368 | 80 | ||
More Information
Set Membership: |
|

KIF2C
Genomic Map