This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj05935965_m1
See other MON1B GE Assays ›
Gene Symbol:
MON1B
Gene Name:
MON1 homolog B, secretory trafficking associated
Gene Aliases:
-
Chromosome Location:
Chr.20: 31168380 - 31182209 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj05935965_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MON1B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008986253.2 XP_008984501.1 5 - 6 1790 81
XM_008986254.2 XP_008984502.1 5 - 6 1764 81
XM_008986255.2 XP_008984503.1 5 - 6 1740 81
XM_008986256.2 XP_008984504.1 5 - 6 1581 81
XM_008986257.2 XP_008984505.1 5 - 6 3051 81
XM_008986258.2 XP_008984506.1 5 - 6 1704 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gqkn5:80/100.66.76.113:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0