This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj05945327_m1
See other MS4A6A GE Assays ›
Gene Symbol:
MS4A6A
Gene Name:
membrane spanning 4-domains A6A
Gene Aliases:
-
Chromosome Location:
Chr.11: 115605274 - 115625644 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj05945327_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MS4A6A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002755374.4 XP_002755420.1 5 - 6 912 86
XM_009008234.2 XP_009006482.1 5 - 6 946 86
XM_009008235.2 XP_009006483.1 5 - 6 781 86
XM_009008236.2 XP_009006484.1 5 - 6 864 86
XM_009008237.2 XP_009006485.1 5 - 6 740 86
XM_017978164.1 XP_017833653.1 5 - 6 601 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Probe spans exons

Panther Classification:

Molecular Function -

transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0