This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj05945328_m1
See other MS4A6A GE Assays ›
Gene Symbol:
MS4A6A
Gene Name:
membrane spanning 4-domains A6A
Gene Aliases:
-
Chromosome Location:
Chr.11: 115605274 - 115625644 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj05945328_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MS4A6A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002755374.4 XP_002755420.1 6 - 7 1010 81
XM_009008234.2 XP_009006482.1 6 - 7 1044 81
XM_009008235.2 XP_009006483.1 6 - 7 879 81
XM_009008236.2 XP_009006484.1 6 - 7 962 81
XM_009008237.2 XP_009006485.1 6 - 7 838 81
XM_017978164.1 XP_017833653.1 6 - 7 699 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Probe spans exons

Panther Classification:

Molecular Function -

transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0