This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj05945988_g1
See other RBM39 GE Assays ›
Gene Symbol:
RBM39
Gene Name:
RNA binding motif protein 39
Gene Aliases:
-
Chromosome Location:
Chr.5: 1461218 - 1504539 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj05945988_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RBM39
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002747232.4 XP_002747278.1 11 - 12 1485 76
XM_002747233.4 XP_002747279.1 11 - 12 1489 76
XM_002747234.4 XP_002747280.1 10 - 11 1419 76
XM_002747235.4 XP_002747281.1 10 - 11 1423 76
XM_017971788.1 XP_017827277.1 12 - 13 2181 76
XM_017971789.1 XP_017827278.1 12 - 13 2038 76
XM_017971790.1 XP_017827279.1 11 - 12 1930 76
XM_017971791.1 XP_017827280.1 12 - 13 1562 76
XM_017971792.1 XP_017827281.1 13 - 14 2253 76
XM_017971793.1 XP_017827282.1 11 - 12 2115 76
XM_017971794.1 XP_017827283.1 12 - 13 1561 76
XM_017971795.1 XP_017827284.1 13 - 14 1634 76
XM_017971796.1 XP_017827285.1 11 - 12 1492 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

RNA splicing factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0