This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj05952294_m1
See other ECHDC2 GE Assays ›
Gene Symbol:
ECHDC2
Gene Name:
enoyl-CoA hydratase domain containing 2
Gene Aliases:
-
Chromosome Location:
Chr.7: 88108838 - 88148044 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj05952294_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ECHDC2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002750836.3 XP_002750882.1 3 - 4 735 77
XM_002750837.3 XP_002750883.1 3 - 4 733 77
XM_003733447.3 XP_003733495.1 3 - 4 739 77
XM_009001247.2 XP_008999495.1 3 - 4 747 77
XM_009001248.2 XP_008999496.1 3 - 4 741 77
XM_009001250.2 XP_008999498.1 3 - 4 750 77
XM_009001251.2 XP_008999499.1 3 - 4 745 77
XM_009001252.2 XP_008999500.1 3 - 4 747 77
XM_009001253.2 XP_008999501.1 3 - 4 749 77
XM_017974754.1 XP_017830243.1 3 - 4 737 77
XM_017974755.1 XP_017830244.1 3 - 4 752 77
XM_017974756.1 XP_017830245.1 4 - 5 365 77
XM_017974757.1 XP_017830246.1 3 - 4 733 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

hydratase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0