This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj05957574_m1
See other MS4A7 GE Assays ›
Gene Symbol:
MS4A7
Gene Name:
membrane spanning 4-domains A7
Gene Aliases:
-
Chromosome Location:
Chr.11: 115764962 - 115781937 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj05957574_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MS4A7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002755375.4 XP_002755421.1 4 - 5 554 89
XM_009008241.2 XP_009006489.1 4 - 5 577 89
XM_009008242.2 XP_009006490.1 6 - 7 674 89
XM_009008244.2 XP_009006492.1 3 - 4 420 89
XM_017978168.1 XP_017833657.1 7 - 8 856 89
XM_017978169.1 XP_017833658.1 6 - 7 815 89
XM_017978170.1 XP_017833659.1 5 - 6 633 89
XM_017978171.1 XP_017833660.1 5 - 6 760 89
XM_017978172.1 XP_017833661.1 7 - 8 827 89
XM_017978173.1 XP_017833662.1 6 - 7 749 89
XM_017978174.1 XP_017833663.1 3 - 4 468 89
XM_017978175.1 XP_017833664.1 6 - 7 697 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0