This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj05961886_g1
See other TP53I11 GE Assays ›
Gene Symbol:
TP53I11
Gene Name:
tumor protein p53 inducible protein 11
Gene Aliases:
-
Chromosome Location:
Chr.11: 108131528 - 108149496 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj05961886_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 417.00
Your Price
Online offer: 387.65
387.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TP53I11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_009007977.2 XP_009006225.1 7 - 8 709 83
XM_009007978.2 XP_009006226.1 8 - 9 897 83
XM_009007979.2 XP_009006227.1 7 - 8 699 83
XM_009007980.2 XP_009006228.1 7 - 8 649 83
XM_017978040.1 XP_017833529.1 8 - 9 879 83
XM_017978041.1 XP_017833530.1 7 - 8 626 83
XM_017978042.1 XP_017833531.1 4 - 5 212 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-x666z:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline