This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj05969565_g1
See other TMEM25 GE Assays ›
Gene Symbol:
TMEM25
Gene Name:
transmembrane protein 25
Gene Aliases:
-
Chromosome Location:
Chr.11: 16140014 - 16144843 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj05969565_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMEM25
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002754488.4 XP_002754534.1 5 - 6 926 58
XM_009006914.2 XP_009005162.1 6 - 7 883 58
XM_009006917.2 XP_009005165.1 6 - 7 1041 58
XM_009006918.2 XP_009005166.1 6 - 7 1058 58
XM_009006919.2 XP_009005167.1 6 - 7 1041 58
XM_009006920.2 XP_009005168.1 5 - 6 909 58
XM_017977572.1 XP_017833061.1 5 - 6 1458 58
XM_017977573.1 XP_017833062.1 5 - 6 1458 58
XM_017977574.1 XP_017833063.1 4 - 5 1326 58
XM_017977575.1 XP_017833064.1 5 - 6 1175 58
XM_017977576.1 XP_017833065.1 5 - 6 1175 58
XM_017977577.1 XP_017833066.1 4 - 5 1043 58
XM_017977578.1 XP_017833067.1 4 - 5 1043 58
XM_017977579.1 XP_017833068.1 5 - 6 1175 58

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0