This product has been added to your favorites list. Go to
My Favorites
Please enter the information below and press OK to send your cart to Core Services for purchase.
Name
Phone number
Email
Budget number
PI Name
PI Email
Building and Room number
Pickup
Delivery
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Cj05978301_m1
See other NAT9 GE Assays ›
Gene Symbol:
NAT9
Gene Name:
N-acetyltransferase 9 (putative)
Gene Aliases:
-
Chromosome Location:
Chr.5: 123985195 - 123990305 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID
Cj05978301_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: NAT9
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_008997714.2
XP_008995962.2
5 - 6
509
76
XM_008997715.2
XP_008995963.2
5 - 6
505
76
XM_008997716.2
XP_008995964.1
4 - 5
482
76
XM_008997718.2
XP_008995966.1
4 - 5
485
76
XM_008997719.2
XP_008995967.1
5 - 6
430
76
XM_008997720.2
XP_008995968.1
4 - 5
482
76
XM_017972800.1
XP_017828289.1
5 - 6
510
76
XM_017972801.1
XP_017828290.1
5 - 6
506
76
XM_017972802.1
XP_017828291.1
5 - 6
518
76
XM_017972803.1
XP_017828292.1
4 - 5
434
76
XM_017972804.1
XP_017828293.1
4 - 5
424
76
XM_017972805.1
XP_017828294.1
4 - 5
419
76
XM_017972806.1
XP_017828295.1
4 - 5
647
76
XM_017972807.1
XP_017828296.1
4 - 5
636
76
XM_017972808.1
XP_017828297.1
5 - 6
518
76
XM_017972809.1
XP_017828298.1
4 - 5
657
76
XM_017972810.1
XP_017828299.1
5 - 6
508
76
XM_017972811.1
XP_017828300.1
4 - 5
820
76
XM_017972812.1
XP_017828301.1
4 - 5
485
76
XM_017972813.1
XP_017828302.1
4 - 5
482
76
XM_017972814.1
XP_017828303.1
4 - 5
695
76
XM_017972815.1
XP_017828304.1
4 - 5
482
76
XM_017972816.1
XP_017828305.1
4 - 5
534
76
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
3' Most
Recommended Assay
Probe spans exons
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0