This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj05980663_g1
See other RPE GE Assays ›
Gene Symbol:
RPE
Gene Name:
ribulose-5-phosphate-3-epimerase
Gene Aliases:
-
Chromosome Location:
Chr.6: 126144345 - 126159904 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj05980663_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPE
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002749737.4 XP_002749783.1 4 - 5 576 93
XM_008999406.2 XP_008997654.1 4 - 5 474 93
XM_008999408.2 XP_008997656.1 4 - 5 495 93
XM_008999409.2 XP_008997657.1 5 - 6 543 93
XM_008999410.2 XP_008997658.1 5 - 6 522 93
XM_008999412.2 XP_008997660.1 3 - 4 442 93
XM_008999413.2 XP_008997661.1 4 - 5 448 93
XM_008999414.2 XP_008997662.1 3 - 4 411 93
XM_008999415.2 XP_008997663.1 4 - 5 576 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-lqq8t:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0