Assay Details
| Gene Symbol: MAP3K21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003735282.3 | XP_003735330.1 | 8 - 9 | 2135 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAP3K21| Gene Symbol: MAP3K21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003735282.3 | XP_003735330.1 | 8 - 9 | 2135 | 81 | |
Set Membership: |
|