This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj05985552_m1
See other BAAT GE Assays ›
Gene Symbol:
BAAT
Gene Name:
bile acid-CoA:amino acid N-acyltransferase
Gene Aliases:
-
Chromosome Location:
Chr.1: 144523485 - 144553664 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj05985552_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BAAT
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002743159.4 XP_002743205.2 3 - 4 2323 77
XM_009000359.2 XP_008998607.1 4 - 5 2414 77
XM_009000367.2 XP_008998615.1 5 - 6 2506 77
XM_017975069.1 XP_017830558.1 3 - 4 1054 77
XM_017975072.1 XP_017830561.1 4 - 5 1110 77
XM_017975073.1 XP_017830562.1 4 - 5 1142 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

esterase hydrolase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0