This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj05987206_m1
See other POLR3D GE Assays ›
Gene Symbol:
POLR3D
Gene Name:
RNA polymerase III subunit D
Gene Aliases:
-
Chromosome Location:
Chr.13: 15960503 - 15968869 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj05987206_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: POLR3D
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017963700.1 XP_017819189.1 1 - 2 365 99
XM_017963701.1 XP_017819190.1 2 - 3 1163 99
XM_017963702.1 XP_017819191.1 1 - 2 365 99
XM_017963703.1 XP_017819192.1 1 - 2 365 99
XM_017963704.1 XP_017819193.1 1 - 2 365 99

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7gdsv:80/100.66.79.88:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0