This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj05989906_m1
See other IGSF10 GE Assays ›
Gene Symbol:
IGSF10
Gene Name:
immunoglobulin superfamily member 10
Gene Aliases:
-
Chromosome Location:
Chr.17: 25631582 - 25726746 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj05989906_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IGSF10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002759487.3 XP_002759533.1 6 - 7 5351 78
XM_017965748.1 XP_017821237.1 10 - 11 6547 78
XM_017965749.1 XP_017821238.1 8 - 9 7062 78
XM_017965750.1 XP_017821239.1 11 - 12 6020 78
XM_017965751.1 XP_017821240.1 11 - 12 6001 78
XM_017965752.1 XP_017821241.1 8 - 9 6246 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

immunoglobulin superfamily cell adhesion molecule

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline