Assay Details
| Gene Symbol: ALDH3A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008996754.2 | XP_008995002.1 | 10 - 11 | 1273 | 69 | |
| XM_008996755.2 | XP_008995003.1 | 9 - 10 | 1179 | 69 | ||
| XM_008996756.2 | XP_008995004.1 | 7 - 8 | 1043 | 69 | ||
| XM_008996758.2 | XP_008995006.1 | 10 - 11 | 1273 | 69 | ||
More Information
Set Membership: |
|

ALDH3A2
Genomic Map