This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj05991580_g1
See other MSANTD4 GE Assays ›
Gene Symbol:
MSANTD4
Gene Name:
Myb/SANT DNA binding domain containing 4 with coiled-coils
Gene Aliases:
-
Chromosome Location:
Chr.11: 3497290 - 3510211 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj05991580_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MSANTD4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_009006715.2 XP_009004963.1 1 - 2 3221 83
XM_009006716.2 XP_009004964.1 3 - 4 753 83
XM_009006717.2 XP_009004965.1 2 - 3 816 83
XM_009006718.2 XP_009004966.1 2 - 3 1056 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0