This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj05993967_m1
See other HSD17B7 GE Assays ›
Gene Symbol:
HSD17B7
Gene Name:
hydroxysteroid 17-beta dehydrogenase 7
Gene Aliases:
-
Chromosome Location:
Chr.18: 25532602 - 25561359 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj05993967_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HSD17B7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002807706.4 XP_002807752.2 2 - 3 340 95
XM_008984909.2 XP_008983157.1 2 - 3 234 95
XM_017966157.1 XP_017821646.1 2 - 3 340 95
XM_017966158.1 XP_017821647.1 2 - 3 340 95
XM_017966159.1 XP_017821648.1 2 - 3 340 95
GenBank mRNA AF263468.1 - 1 - 2 212 95

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

reductase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-5c659bcd8-r6ctg:80/100.66.75.22:80.
git-commit: 653a2953ec6bb2e3dd1e37289b36fc4e0ab15bfe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-Offline