This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj06002235_m1
See other DTX3 GE Assays ›
Gene Symbol:
DTX3
Gene Name:
deltex E3 ubiquitin ligase 3
Gene Aliases:
-
Chromosome Location:
Chr.9: 46435658 - 46441385 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj06002235_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DTX3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_009004106.2 XP_009002354.1 3 - 4 1581 72
XM_009004107.2 XP_009002355.1 4 - 5 985 72
XM_009004108.2 XP_009002356.1 3 - 4 1599 72
XM_009004109.2 XP_009002357.1 4 - 5 1439 72
XM_009004110.2 XP_009002358.1 3 - 4 1576 72
XM_009004111.2 XP_009002359.1 3 - 4 1594 72
XM_017976162.1 XP_017831651.1 4 - 5 1421 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-5thf6:80/100.66.79.225:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0