Assay Details
| Gene Symbol: GSTZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754151.4 | XP_002754197.1 | 6 - 7 | 720 | 59 | |
| XM_003734049.3 | XP_003734097.1 | 5 - 6 | 594 | 59 | ||
| XM_009006385.2 | XP_009004633.1 | 7 - 8 | 699 | 59 | ||
| XM_017977246.1 | XP_017832735.1 | 7 - 8 | 702 | 59 | ||
| XM_017977247.1 | XP_017832736.1 | 6 - 7 | 720 | 59 | ||
| XM_017977248.1 | XP_017832737.1 | 6 - 7 | 591 | 59 | ||
| XM_017977249.1 | XP_017832738.1 | 6 - 7 | 575 | 59 | ||
More Information
Set Membership: |
|

GSTZ1
Genomic Map