This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj06017049_m1
See other SLC7A6 GE Assays ›
Gene Symbol:
SLC7A6
Gene Name:
solute carrier family 7 member 6
Gene Aliases:
-
Chromosome Location:
Chr.20: 22064217 - 22105927 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj06017049_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC7A6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002761086.4 XP_002761132.1 3 - 4 716 68
XM_008986112.2 XP_008984360.1 5 - 6 1120 68
XM_008986114.2 XP_008984362.1 5 - 6 921 68
XM_008986115.2 XP_008984363.1 5 - 6 1954 68
XM_008986117.2 XP_008984365.1 4 - 5 880 68
XM_008986118.2 XP_008984366.1 4 - 5 1044 68
XM_008986119.2 XP_008984367.1 5 - 6 1051 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline