This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj06017394_m1
See other ELAVL2 GE Assays ›
Gene Symbol:
ELAVL2
Gene Name:
ELAV like RNA binding protein 2
Gene Aliases:
-
Chromosome Location:
Chr.1: 125480100 - 125618608 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj06017394_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ELAVL2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002742980.4 XP_002743026.1 2 - 3 784 85
XM_002742981.3 XP_002743027.1 3 - 4 492 85
XM_008997795.2 XP_008996043.1 3 - 4 875 85
XM_008997821.2 XP_008996069.1 2 - 3 784 85
XM_017973917.1 XP_017829406.1 3 - 4 900 85
XM_017973922.1 XP_017829411.1 3 - 4 900 85
XM_017973924.1 XP_017829413.1 3 - 4 875 85
XM_017973925.1 XP_017829414.1 3 - 4 872 85
XM_017973929.1 XP_017829418.1 3 - 4 900 85
XM_017973930.1 XP_017829419.1 3 - 4 900 85
XM_017973934.1 XP_017829423.1 3 - 4 875 85
XM_017973941.1 XP_017829430.1 3 - 4 900 85

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

translation initiation factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0